Hexafluoroisopropyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450338-25G
APOSPC450338-25GEA 1540 EUR
APOSPC450338-25G
Hexafluoroisopropyl 1H,1H-perfluorohexyl carbonate
Hexafluoroisopropyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₀H₃F₁₇O₃
Molecular Weight: 494,102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334331
CAS-Nummer: 1980048-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR