2,2,3,3,3-Pentafluoropropyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450339-25G
APOSPC450339-25GEA
1540
EUR
APOSPC450339-25G
2,2,3,3,3-Pentafluoropropyl 1H,1H-perfluorohexyl carbonate
2,2,3,3,3-Pentafluoropropyl 1H,1H-perfluorohexyl carbonate
Formel:
C₁₀H₄F₁₆O₃ Molecular Weight: 476.111 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334332 CAS-Nummer: 1980076-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...