1H,1H,7H-Perfluoroheptyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450341-25G
APOSPC450341-25GEA 1540 EUR
APOSPC450341-25G
1H,1H,7H-Perfluoroheptyl 2,2,2-trifluoroethyl carbonate
1H,1H,7H-Perfluoroheptyl 2,2,2-trifluoroethyl carbonate
Formel: C₁₀H₅F₁₅O₃
Molecular Weight: 458,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334334
CAS-Nummer: 1980044-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR