Hexafluoroisopropyl 1H,1H,2H,2H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450342-25G
APOSPC450342-25GEA
1540
EUR
APOSPC450342-25G
Hexafluoroisopropyl 1H,1H,2H,2H-perfluorohexyl carbonate
Hexafluoroisopropyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel:
C₁₀H₅F₁₅O₃ Molecular Weight: 458,121 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334335 CAS-Nummer: 1980076-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...