2,2-Difluoroethyl 1H,1H,7H-perfluoroheptyl carbonate

Lieferant: Apollo Scientific

PC450343-25G
APOSPC450343-25GEA 1540 EUR
APOSPC450343-25G
2,2-Difluoroethyl 1H,1H,7H-perfluoroheptyl carbonate
2,2-Difluoroethyl 1H,1H,7H-perfluoroheptyl carbonate
Formel: C₁₀H₆F₁₄O₃
Molecular Weight: 440,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334336
CAS-Nummer: 1980035-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR