1H,1H,2H,2H-Perfluorohexyl 2,2,3,3,3-pentafluoropropyl carbonate
Lieferant: Apollo Scientific
PC450344-25G
APOSPC450344-25GEA
1540
EUR
APOSPC450344-25G
1H,1H,2H,2H-Perfluorohexyl 2,2,3,3,3-pentafluoropropyl carbonate
1H,1H,2H,2H-Perfluorohexyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel:
C₁₀H₆F₁₄O₃ Molecular Weight: 440,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334337 CAS-Nummer: 1980048-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...