1H,1H,2H,2H-Perfluorohexyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450345-25G
APOSPC450345-25GEA 1540 EUR
APOSPC450345-25G
1H,1H,2H,2H-Perfluorohexyl 2,2,3,3-tetrafluoropropyl carbonate
1H,1H,2H,2H-Perfluorohexyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₁₀H₇F₁₃O₃
Molecular Weight: 422,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334338
CAS-Nummer: 1980044-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR