3-Fluoropropyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450346-25G
APOSPC450346-25GEA 1540 EUR
APOSPC450346-25G
3-Fluoropropyl 1H,1H-perfluorohexyl carbonate
3-Fluoropropyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₀H₈F₁₂O₃
Molecular Weight: 404,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334339
CAS-Nummer: 1980085-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR