2,2,3,3,4,4,4-Heptafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450347-25G
APOSPC450347-25GEA 1540 EUR
APOSPC450347-25G
2,2,3,3,4,4,4-Heptafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₀H₈F₁₂O₃
Molecular Weight: 404,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334340
CAS-Nummer: 1980048-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR