2,2,3,4,4,4-Hexafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450348-25G
APOSPC450348-25GEA 1540 EUR
APOSPC450348-25G
2,2,3,4,4,4-Hexafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate
2,2,3,4,4,4-Hexafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₀H₉F₁₁O₃
Molecular Weight: 386,159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334341
CAS-Nummer: 1980086-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR