3-Fluoropropyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450349-25G
APOSPC450349-25GEA 1540 EUR
APOSPC450349-25G
3-Fluoropropyl 1H,1H,2H,2H-perfluorohexyl carbonate
3-Fluoropropyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₀H₁₀F₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334342
CAS-Nummer: 1980085-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR