2,2,3,3,4,4,4-Heptafluorobutyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450350-25G
APOSPC450350-25GEA 1540 EUR
APOSPC450350-25G
2,2,3,3,4,4,4-Heptafluorobutyl 1H,1H-perfluorohexyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₁H₄F₁₈O₃
Molecular Weight: 526,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334343
CAS-Nummer: 1980039-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR