1H,1H,7H-Perfluoroheptyl hexafluoroisopropyl carbonate
Lieferant: Apollo Scientific
PC450351-25G
APOSPC450351-25GEA
1540
EUR
APOSPC450351-25G
1H,1H,7H-Perfluoroheptyl hexafluoroisopropyl carbonate
1H,1H,7H-Perfluoroheptyl hexafluoroisopropyl carbonate
Formel:
C₁₁H₄F₁₈O₃ Molecular Weight: 526,119 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334344 CAS-Nummer: 1980048-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...