2,2,3,4,4,4-Hexafluorobutyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450352-25G
APOSPC450352-25GEA 1540 EUR
APOSPC450352-25G
2,2,3,4,4,4-Hexafluorobutyl 1H,1H-perfluorohexyl carbonate
2,2,3,4,4,4-Hexafluorobutyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₁H₅F₁₇O₃
Molecular Weight: 508,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334345
CAS-Nummer: 1980086-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR