1H,1H,7H-Perfluoroheptyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450353-25G
APOSPC450353-25GEA 1540 EUR
APOSPC450353-25G
1H,1H,7H-Perfluoroheptyl 2,2,3,3,3-pentafluoropropyl carbonate
1H,1H,7H-Perfluoroheptyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₁₁H₅F₁₇O₃
Molecular Weight: 508,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334346
CAS-Nummer: 1980034-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR