2,2,3,3,4,4,4-Heptafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450354-25G
APOSPC450354-25GEA 1540 EUR
APOSPC450354-25G
2,2,3,3,4,4,4-Heptafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₁H₆F₁₆O₃
Molecular Weight: 490,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334347
CAS-Nummer: 1980045-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR