1H,1H,7H-Perfluoroheptyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450355-25G
APOSPC450355-25GEA 1540 EUR
APOSPC450355-25G
1H,1H,7H-Perfluoroheptyl 2,2,3,3-tetrafluoropropyl carbonate
1H,1H,7H-Perfluoroheptyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₁₁H₆F₁₆O₃
Molecular Weight: 490,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334348
CAS-Nummer: 1980038-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR