2,2,3,4,4,4-Hexafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450356-25G
APOSPC450356-25GEA 1540 EUR
APOSPC450356-25G
2,2,3,4,4,4-Hexafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
2,2,3,4,4,4-Hexafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₁H₇F₁₅O₃
Molecular Weight: 472,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334349
CAS-Nummer: 1980085-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR