1H,1H,7H-Perfluoroheptyl 3-fluoropropyl carbonate

Lieferant: Apollo Scientific

PC450357-25G
APOSPC450357-25GEA 1540 EUR
APOSPC450357-25G
1H,1H,7H-Perfluoroheptyl 3-fluoropropyl carbonate
1H,1H,7H-Perfluoroheptyl 3-fluoropropyl carbonate
Formel: C₁₁H₉F₁₃O₃
Molecular Weight: 436,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334350
CAS-Nummer: 1980085-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR