Bis(4,4,5,5,5-pentafluoropentyl) carbonate
Lieferant: Apollo Scientific
PC450358-25G
APOSPC450358-25GEA
1540
EUR
APOSPC450358-25G
Bis(4,4,5,5,5-pentafluoropentyl) carbonate
Bis(4,4,5,5,5-pentafluoropentyl) carbonate
Formel:
C₁₁H₁₂F₁₀O₃ Molecular Weight: 382,195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334351 CAS-Nummer: 1980048-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...