1H,1H,9H-Perfluorononyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450359-25G
APOSPC450359-25GEA 1540 EUR
APOSPC450359-25G
1H,1H,9H-Perfluorononyl 2,2,2-trifluoroethyl carbonate
1H,1H,9H-Perfluorononyl 2,2,2-trifluoroethyl carbonate
Formel: C₁₂H₅F₁₉O₃
Molecular Weight: 558,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334352
CAS-Nummer: 1980035-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR