1H,1H,7H-Perfluoroheptyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Lieferant: Apollo Scientific
PC450360-25G
APOSPC450360-25GEA
1540
EUR
APOSPC450360-25G
1H,1H,7H-Perfluoroheptyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
1H,1H,7H-Perfluoroheptyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formel:
C₁₂H₅F₁₉O₃ Molecular Weight: 558,136 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334353 CAS-Nummer: 1980086-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...