2,2-Difluoroethyl 1H,1H,9H-perfluorononyl carbonate

Lieferant: Apollo Scientific

PC450361-25G
APOSPC450361-25GEA 1540 EUR
APOSPC450361-25G
2,2-Difluoroethyl 1H,1H,9H-perfluorononyl carbonate
2,2-Difluoroethyl 1H,1H,9H-perfluorononyl carbonate
Formel: C₁₂H₆F₁₈O₃
Molecular Weight: 540,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334354
CAS-Nummer: 1980076-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR