1H,1H,7H-Perfluoroheptyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450362-25G
APOSPC450362-25GEA 1540 EUR
APOSPC450362-25G
1H,1H,7H-Perfluoroheptyl 2,2,3,4,4,4-hexafluorobutyl carbonate
1H,1H,7H-Perfluoroheptyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel: C₁₂H₆F₁₈O₃
Molecular Weight: 540,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334355
CAS-Nummer: 1980045-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR