4,4,5,5,5-Pentafluoropentyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450363-25G
APOSPC450363-25GEA 1540 EUR
APOSPC450363-25G
4,4,5,5,5-Pentafluoropentyl 1H,1H-perfluorohexyl carbonate
4,4,5,5,5-Pentafluoropentyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₂H₈F₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334356
CAS-Nummer: 1980034-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR