1H,1H,2H,2H-Perfluorohexyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450364-25G
APOSPC450364-25GEA 1540 EUR
APOSPC450364-25G
1H,1H,2H,2H-Perfluorohexyl 4,4,5,5,5-pentafluoropentyl carbonate
1H,1H,2H,2H-Perfluorohexyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₂H₁₀F₁₄O₃
Molecular Weight: 468,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334357
CAS-Nummer: 1980086-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR