Bis(1H,1H-perfluorohexyl) carbonate

Lieferant: Apollo Scientific

PC450365-25G
APOSPC450365-25GEA 1540 EUR
APOSPC450365-25G
Bis(1H,1H-perfluorohexyl) carbonate
Bis(1H,1H-perfluorohexyl) carbonate
Formel: C₁₃H₄F₂₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334358
CAS-Nummer: 1980085-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR