1H,1H,9H-Perfluorononyl hexafluoroisopropyl carbonate

Lieferant: Apollo Scientific

PC450366-25G
APOSPC450366-25GEA 1540 EUR
APOSPC450366-25G
1H,1H,9H-Perfluorononyl hexafluoroisopropyl carbonate
1H,1H,9H-Perfluorononyl hexafluoroisopropyl carbonate
Formel: C₁₃H₄F₂₂O₃
Molecular Weight: 626,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334359
CAS-Nummer: 1980048-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR