1H,1H,9H-Perfluorononyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450367-25G
APOSPC450367-25GEA 1540 EUR
APOSPC450367-25G
1H,1H,9H-Perfluorononyl 2,2,3,3,3-pentafluoropropyl carbonate
1H,1H,9H-Perfluorononyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₁₃H₅F₂₁O₃
Molecular Weight: 608.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334360
CAS-Nummer: 1980086-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR