1H,1H,9H-Perfluorononyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450368-25G
APOSPC450368-25GEA 1540 EUR
APOSPC450368-25G
1H,1H,9H-Perfluorononyl 2,2,3,3-tetrafluoropropyl carbonate
1H,1H,9H-Perfluorononyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₁₃H₆F₂₀O₃
Molecular Weight: 590,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334361
CAS-Nummer: 1980034-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR