1H,1H,2H,2H-Perfluorohexyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450369-25G
APOSPC450369-25GEA
1540
EUR
APOSPC450369-25G
1H,1H,2H,2H-Perfluorohexyl 1H,1H-perfluorohexyl carbonate
1H,1H,2H,2H-Perfluorohexyl 1H,1H-perfluorohexyl carbonate
Formel:
C₁₃H₆F₂₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334362 CAS-Nummer: 1980085-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...