Bis(1H,1H,2H,2H-perfluorohexyl) carbonate

Lieferant: Apollo Scientific

PC450370-25G
APOSPC450370-25GEA 1540 EUR
APOSPC450370-25G
Bis(1H,1H,2H,2H-perfluorohexyl) carbonate
Bis(1H,1H,2H,2H-perfluorohexyl) carbonate
Formel: C₁₃H₈F₁₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334363
CAS-Nummer: 1132677-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR