3-Fluoropropyl 1H,1H,9H-perfluorononyl carbonate

Lieferant: Apollo Scientific

PC450371-25G
APOSPC450371-25GEA 1540 EUR
APOSPC450371-25G
3-Fluoropropyl 1H,1H,9H-perfluorononyl carbonate
3-Fluoropropyl 1H,1H,9H-perfluorononyl carbonate
Formel: C₁₃H₉F₁₇O₃
Molecular Weight: 536,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334364
CAS-Nummer: 1980034-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR