1H,1H,7H-Perfluoroheptyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450372-25G
APOSPC450372-25GEA 1540 EUR
APOSPC450372-25G
1H,1H,7H-Perfluoroheptyl 4,4,5,5,5-pentafluoropentyl carbonate
1H,1H,7H-Perfluoroheptyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₃H₉F₁₇O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334365
CAS-Nummer: 1980034-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR