1H,1H,7H-Perfluoroheptyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450374-25G
APOSPC450374-25GEA
1540
EUR
APOSPC450374-25G
1H,1H,7H-Perfluoroheptyl 1H,1H-perfluorohexyl carbonate
1H,1H,7H-Perfluoroheptyl 1H,1H-perfluorohexyl carbonate
Formel:
C₁₄H₅F₂₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334367 CAS-Nummer: 1980034-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...