1H,1H,9H-Perfluorononyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450375-25G
APOSPC450375-25GEA 1540 EUR
APOSPC450375-25G
1H,1H,9H-Perfluorononyl 2,2,3,4,4,4-hexafluorobutyl carbonate
1H,1H,9H-Perfluorononyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel: C₁₄H₆F₂₂O₃
Molecular Weight: 640,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334368
CAS-Nummer: 1980034-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR