1H,1H,7H-Perfluoroheptyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450376-25G
APOSPC450376-25GEA 1540 EUR
APOSPC450376-25G
1H,1H,7H-Perfluoroheptyl 1H,1H,2H,2H-perfluorohexyl carbonate
1H,1H,7H-Perfluoroheptyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₄H₇F₂₁O₃
Molecular Weight: 622,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334369
CAS-Nummer: 1980085-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR