1H,1H,9H-Perfluorononyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450378-25G
APOSPC450378-25GEA 1540 EUR
APOSPC450378-25G
1H,1H,9H-Perfluorononyl 4,4,5,5,5-pentafluoropentyl carbonate
1H,1H,9H-Perfluorononyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₅H₉F₂₁O₃
Molecular Weight: 636,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334370
CAS-Nummer: 1980045-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR