1H,1H,9H-Perfluorononyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450379-25G
APOSPC450379-25GEA 1540 EUR
APOSPC450379-25G
1H,1H,9H-Perfluorononyl 1H,1H-perfluorohexyl carbonate
1H,1H,9H-Perfluorononyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₆H₅F₂₇O₃
Molecular Weight: 758,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334371
CAS-Nummer: 1980085-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR