1H,1H,9H-Perfluorononyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450380-25G
APOSPC450380-25GEA 1540 EUR
APOSPC450380-25G
1H,1H,9H-Perfluorononyl 1H,1H,2H,2H-perfluorohexyl carbonate
1H,1H,9H-Perfluorononyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₆H₇F₂₅O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334372
CAS-Nummer: 1980039-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR