1H,1H,7H-Perfluoroheptyl 1H,1H,9H-perfluorononyl carbonate

Lieferant: Apollo Scientific

PC450381-25G
APOSPC450381-25GEA 1540 EUR
APOSPC450381-25G
1H,1H,7H-Perfluoroheptyl 1H,1H,9H-perfluorononyl carbonate
1H,1H,7H-Perfluoroheptyl 1H,1H,9H-perfluorononyl carbonate
Formel: C₁₇H₆F₂₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334373
CAS-Nummer: 1980034-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR