Bis(1H,1H,9H-perfluorononyl) carbonate
Lieferant: Apollo Scientific
PC450382-25G
APOSPC450382-25GEA
1540
EUR
APOSPC450382-25G
Bis(1H,1H,9H-perfluorononyl) carbonate
Bis(1H,1H,9H-perfluorononyl) carbonate
Formel:
C₁₉H₆F₃₂O₃ Molecular Weight: 890,198 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334374 CAS-Nummer: 2264-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...