4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450383-25G
APOSPC450383-25GEA 1540 EUR
APOSPC450383-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,2-trifluoroethyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,2-trifluoroethyl carbonate
Formel: C₇H₆BrF₇O₃
Molecular Weight: 351,014 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334455
CAS-Nummer: 1980035-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR