4-Bromo-3,3,4,4-tetrafluorobutyl 2,2-difluoroethyl carbonate

Lieferant: Apollo Scientific

PC450384-25G
APOSPC450384-25GEA 1540 EUR
APOSPC450384-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2-difluoroethyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2-difluoroethyl carbonate
Formel: C₇H₇BrF₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334456
CAS-Nummer: 1980034-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR