4-Bromo-3,3,4,4-tetrafluorobutyl hexafluoroisopropyl carbonate

Lieferant: Apollo Scientific

PC450385-25G
APOSPC450385-25GEA 1540 EUR
APOSPC450385-25G
4-Bromo-3,3,4,4-tetrafluorobutyl hexafluoroisopropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl hexafluoroisopropyl carbonate
Formel: C₈H₅BrF₁₀O₃
Molecular Weight: 419,012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334457
CAS-Nummer: 1980085-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR