4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450386-25G
APOSPC450386-25GEA 1540 EUR
APOSPC450386-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₈H₆BrF₉O₃
Molecular Weight: 401.021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334458
CAS-Nummer: 1923267-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR