4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
Lieferant: Apollo Scientific
PC450387-25G
APOSPC450387-25GEA
1540
EUR
APOSPC450387-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
Formel:
C₈H₇BrF₈O₃ Molecular Weight: 383,031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334459 CAS-Nummer: 1980035-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...