4-Bromo-3,3,4,4-tetrafluorobutyl 3-fluoropropyl carbonate

Lieferant: Apollo Scientific

PC450388-25G
APOSPC450388-25GEA 1540 EUR
APOSPC450388-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 3-fluoropropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 3-fluoropropyl carbonate
Formel: C₈H₁₀BrF₅O₃
Molecular Weight: 329,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334460
CAS-Nummer: 1923065-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR