4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Lieferant: Apollo Scientific
PC450390-25G
APOSPC450390-25GEA
1540
EUR
APOSPC450390-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,4,4,4-hexafluorobutyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel:
C₉H₇BrF₁₀O₃ Molecular Weight: 433,038 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334462 CAS-Nummer: 1980034-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...