Bis(4-bromo-3,3,4,4-tetrafluorobutyl) carbonate

Lieferant: Apollo Scientific

PC450391-25G
APOSPC450391-25GEA 1540 EUR
APOSPC450391-25G
Bis(4-bromo-3,3,4,4-tetrafluorobutyl) carbonate
Bis(4-bromo-3,3,4,4-tetrafluorobutyl) carbonate
Formel: C₉H₈Br₂F₈O₃
Molecular Weight: 475,953 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334463
CAS-Nummer: 1923267-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR